Anti-PRKAR1B Mouse Monoclonal Antibody [clone: OTI12E5]
Proveedor: OriGene
ORIGTA502972EA 581 EUR
ORIGTA502972
Anti-PRKAR1B Mouse Monoclonal Antibody [clone: OTI12E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PRKAR1B (protein kinase, cAMP-dependent, regulatory, type I, beta)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human