Anti-AK4 Mouse Monoclonal Antibody [clone: OTI5E7]

Proveedor: OriGene
ORIGTA502945EA 603 EUR
ORIGTA502945
Anti-AK4 Mouse Monoclonal Antibody [clone: OTI5E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK4 (adenylate kinase 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR