Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2E11]

Proveedor: OriGene
ORIGTA502916EA 603 EUR
ORIGTA502916
Anti-HDHD2 Mouse Monoclonal Antibody [clone: OTI2E11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HDHD2 (haloacid dehalogenase-like hydrolase domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR