Anti-EIF4E Mouse Monoclonal Antibody [clone: OTI6C3]

Proveedor: OriGene
TA502896
ORIGTA502896EA 616 EUR
ORIGTA502896
Anti-EIF4E Mouse Monoclonal Antibody [clone: OTI6C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR