Anti-MGLL Mouse Monoclonal Antibody [clone: OTI1H2]

Proveedor: OriGene
ORIGTA502888EA 603 EUR
ORIGTA502888
Anti-MGLL Mouse Monoclonal Antibody [clone: OTI1H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: MGLL (monoglyceride lipase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR