Anti-MAPRE2 Mouse Monoclonal Antibody [clone: OTI7C10]

Proveedor: OriGene
ORIGTA502871EA 603 EUR
ORIGTA502871
Anti-MAPRE2 Mouse Monoclonal Antibody [clone: OTI7C10]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPRE2 (microtubule-associated protein, RP/EB family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR