Anti-PRKAR1B Mouse Monoclonal Antibody [clone: OTI9C5]

Proveedor: OriGene
ORIGTA502793EA 581 EUR
ORIGTA502793
Anti-PRKAR1B Mouse Monoclonal Antibody [clone: OTI9C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKAR1B (protein kinase, cAMP-dependent, regulatory, type I, beta)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR