Anti-LMCD1 Mouse Monoclonal Antibody [clone: OTI2F12]

Proveedor: OriGene
ORIGTA502789EA 603 EUR
ORIGTA502789
Anti-LMCD1 Mouse Monoclonal Antibody [clone: OTI2F12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: LMCD1 (LIM and cysteine-rich domains 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR