Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI6D6]

Proveedor: OriGene
ORIGTA502739EA 603 EUR
ORIGTA502739
Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI6D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR