Anti-PDLIM2 Mouse Monoclonal Antibody [clone: OTI10C4]

Proveedor: OriGene
ORIGTA502708EA 616 EUR
ORIGTA502708
Anti-PDLIM2 Mouse Monoclonal Antibody [clone: OTI10C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PDLIM2 (PDZ and LIM domain 2 (mystique))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR