Anti-PDHA1 Mouse Monoclonal Antibody [clone: OTI2B10]

Proveedor: OriGene
ORIGTA502704EA 608 EUR
ORIGTA502704
Anti-PDHA1 Mouse Monoclonal Antibody [clone: OTI2B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDHA1 (pyruvate dehydrogenase (lipoamide) alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR