Anti-DSTN Mouse Monoclonal Antibody [clone: OTI2E10]

Proveedor: OriGene
ORIGTA502607EA 603 EUR
ORIGTA502607
Anti-DSTN Mouse Monoclonal Antibody [clone: OTI2E10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DSTN (destrin (actin depolymerizing factor))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR