Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI4B10]

Proveedor: OriGene
ORIGTA502566EA 603 EUR
ORIGTA502566
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI4B10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR