Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI3A12]

Proveedor: OriGene
ORIGTA502540EA 603 EUR
ORIGTA502540
Anti-PRKCE Mouse Monoclonal Antibody [clone: OTI3A12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKCE (protein kinase C, epsilon)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR