Anti-NNMT Mouse Monoclonal Antibody [clone: OTI2A3]

Proveedor: OriGene
ORIGTA502538EA 581 EUR
ORIGTA502538
Anti-NNMT Mouse Monoclonal Antibody [clone: OTI2A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NNMT (nicotinamide N-methyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR