Anti-GBA3 Mouse Monoclonal Antibody [clone: OTI5G5]

Proveedor: OriGene
ORIGTA502533EA 603 EUR
ORIGTA502533
Anti-GBA3 Mouse Monoclonal Antibody [clone: OTI5G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GBA3 (glucosidase, beta, acid 3 (gene/pseudogene))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR