Anti-PON1 Mouse Monoclonal Antibody [clone: OTI2D4]

Proveedor: OriGene
ORIGTA502528EA 603 EUR
ORIGTA502528
Anti-PON1 Mouse Monoclonal Antibody [clone: OTI2D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PON1 (paraoxonase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR