Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI2D2]

Proveedor: OriGene
ORIGTA502500EA 603 EUR
ORIGTA502500
Anti-MEF2C Mouse Monoclonal Antibody [clone: OTI2D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MEF2C (myocyte enhancer factor 2C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR