Anti-VASP Mouse Monoclonal Antibody [clone: OTI1F3]

Proveedor: OriGene
ORIGTA502491EA 581 EUR
ORIGTA502491
Anti-VASP Mouse Monoclonal Antibody [clone: OTI1F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR