Anti-UBXN2B Mouse Monoclonal Antibody [clone: OTI2E8]

Proveedor: OriGene
ORIGTA502483EA 581 EUR
ORIGTA502483
Anti-UBXN2B Mouse Monoclonal Antibody [clone: OTI2E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: UBXN2B (UBX domain protein 2B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR