Anti-HAGHL Mouse Monoclonal Antibody [clone: OTI1G8]

Proveedor: OriGene
ORIGTA502476EA 581 EUR
ORIGTA502476
Anti-HAGHL Mouse Monoclonal Antibody [clone: OTI1G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HAGHL (hydroxyacylglutathione hydrolase-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR