Anti-PIH1D2 Mouse Monoclonal Antibody [clone: OTI4A10]

Proveedor: OriGene
ORIGTA502467EA 581 EUR
ORIGTA502467
Anti-PIH1D2 Mouse Monoclonal Antibody [clone: OTI4A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PIH1D2 (PIH1 domain containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR