Anti-KATNAL1 Mouse Monoclonal Antibody [clone: OTI4G1]

Proveedor: OriGene
ORIGTA502459EA 581 EUR
ORIGTA502459
Anti-KATNAL1 Mouse Monoclonal Antibody [clone: OTI4G1]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: KATNAL1 (katanin p60 subunit A-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR