Anti-POLR2J2 Mouse Monoclonal Antibody [clone: OTI3G4]

Proveedor: OriGene
ORIGTA502454EA 603 EUR
ORIGTA502454
Anti-POLR2J2 Mouse Monoclonal Antibody [clone: OTI3G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: POLR2J2 (polymerase (RNA) II (DNA directed) polypeptide J2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR