Anti-HPGD Mouse Monoclonal Antibody [clone: OTI1E9]
Proveedor: OriGene
ORIGTA502452EA 603 EUR
ORIGTA502452
Anti-HPGD Mouse Monoclonal Antibody [clone: OTI1E9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: HPGD (hydroxyprostaglandin dehydrogenase 15-(NAD))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human