Anti-ZFAND2B Mouse Monoclonal Antibody [clone: OTI1G10]

Proveedor: OriGene
ORIGTA502440EA 586 EUR
ORIGTA502440
Anti-ZFAND2B Mouse Monoclonal Antibody [clone: OTI1G10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ZFAND2B (zinc finger, AN1-type domain 2B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR