Anti-VCAM1 Mouse Monoclonal Antibody [clone: OTI3H10]

Proveedor: OriGene
ORIGTA502391EA 608 EUR
ORIGTA502391
Anti-VCAM1 Mouse Monoclonal Antibody [clone: OTI3H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: VCAM1 (vascular cell adhesion molecule 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR