Anti-SERPINB4 Mouse Monoclonal Antibody [clone: OTI4C8]

Proveedor: OriGene
ORIGTA502386EA 603 EUR
ORIGTA502386
Anti-SERPINB4 Mouse Monoclonal Antibody [clone: OTI4C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: SERPINB4 (serpin peptidase inhibitor, clade B (ovalbumin), member 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR