Anti-ZFP36 Mouse Monoclonal Antibody [clone: OTI5E3]

Proveedor: OriGene
ORIGTA502381EA 603 EUR
ORIGTA502381
Anti-ZFP36 Mouse Monoclonal Antibody [clone: OTI5E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ZFP36 (ZFP36 ring finger protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR