Anti-RTFDC1 Mouse Monoclonal Antibody [clone: OTI1E8]

Proveedor: OriGene
ORIGTA502375EA 608 EUR
ORIGTA502375
Anti-RTFDC1 Mouse Monoclonal Antibody [clone: OTI1E8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RTFDC1 (replication termination factor 2 domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR