Anti-DCLRE1B Mouse Monoclonal Antibody [clone: OTI2B7]

Proveedor: OriGene
ORIGTA502352EA 608 EUR
ORIGTA502352
Anti-DCLRE1B Mouse Monoclonal Antibody [clone: OTI2B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DCLRE1B (DNA cross-link repair 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR