Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI1B11]

Proveedor: OriGene
ORIGTA502340EA 603 EUR
ORIGTA502340
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI1B11]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR