Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI9F5]

Proveedor: OriGene
ORIGTA502330EA 603 EUR
ORIGTA502330
Anti-NUDT6 Mouse Monoclonal Antibody [clone: OTI9F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR