Anti-RDH14 Mouse Monoclonal Antibody [clone: OTI3E1]

Proveedor: OriGene
ORIGTA502316EA 603 EUR
ORIGTA502316
Anti-RDH14 Mouse Monoclonal Antibody [clone: OTI3E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: RDH14 (retinol dehydrogenase 14 (all-trans/9-cis/11-cis))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR