Anti-STRIP1 Mouse Monoclonal Antibody [clone: OTI7G7]

Proveedor: OriGene
ORIGTA502314EA 603 EUR
ORIGTA502314
Anti-STRIP1 Mouse Monoclonal Antibody [clone: OTI7G7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STRIP1 (striatin interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR