Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI1H10]

Proveedor: OriGene
ORIGTA502300EA 603 EUR
ORIGTA502300
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: OTI1H10]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR