Anti-ACY3 Mouse Monoclonal Antibody [clone: OTI3A1]

Proveedor: OriGene
ORIGTA502292EA 603 EUR
ORIGTA502292
Anti-ACY3 Mouse Monoclonal Antibody [clone: OTI3A1]
Anticuerpos
Recommended Dilutions: Flow Cytometry: 1:100

Type: Primary
Antigen: ACY3 (aspartoacylase (aminocyclase) 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR