Anti-GBP5 Mouse Monoclonal Antibody [clone: OTI8B12]

Proveedor: OriGene
ORIGTA502286EA 581 EUR
ORIGTA502286
Anti-GBP5 Mouse Monoclonal Antibody [clone: OTI8B12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP5 (guanylate binding protein 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR