Anti-TASP1 Mouse Monoclonal Antibody [clone: OTI1C12]

Proveedor: OriGene
TA502281
ORIGTA502281EA 616 EUR
ORIGTA502281
Anti-TASP1 Mouse Monoclonal Antibody [clone: OTI1C12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: TASP1 (taspase, threonine aspartase, 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR