Anti-MMAA Mouse Monoclonal Antibody [clone: OTI3G5]

Proveedor: OriGene
ORIGTA502277EA 581 EUR
ORIGTA502277
Anti-MMAA Mouse Monoclonal Antibody [clone: OTI3G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MMAA (methylmalonic aciduria (cobalamin deficiency) cblA type)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR