Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI4A5]

Proveedor: OriGene
TA502274
ORIGTA502274EA 593 EUR
ORIGTA502274
Anti-PACSIN3 Mouse Monoclonal Antibody [clone: OTI4A5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PACSIN3 (protein kinase C and casein kinase substrate in neurons 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR