Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI2A10]

Proveedor: OriGene
ORIGTA502252EA 603 EUR
ORIGTA502252
Anti-MSI1 Mouse Monoclonal Antibody [clone: OTI2A10]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MSI1 (musashi RNA-binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR