Anti-BIRC5 Mouse Monoclonal Antibody [clone: OTI6H5]

Proveedor: OriGene
ORIGTA502235EA 603 EUR
ORIGTA502235
Anti-BIRC5 Mouse Monoclonal Antibody [clone: OTI6H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR