Anti-CDCP1 Mouse Monoclonal Antibody [clone: OTI2C1]

Proveedor: OriGene
ORIGTA502228EA 603 EUR
ORIGTA502228
Anti-CDCP1 Mouse Monoclonal Antibody [clone: OTI2C1]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR