Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1D3]

Proveedor: OriGene
ORIGTA502226EA 603 EUR
ORIGTA502226
Anti-CTDSP1 Mouse Monoclonal Antibody [clone: OTI1D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: CTDSP1 (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR