Anti-GPHN Mouse Monoclonal Antibody [clone: OTI3B6]

Proveedor: OriGene
ORIGTA502217EA 608 EUR
ORIGTA502217
Anti-GPHN Mouse Monoclonal Antibody [clone: OTI3B6]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GPHN (gephyrin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR