Anti-MAPK7 Mouse Monoclonal Antibody [clone: OTI1A4]

Proveedor: OriGene
ORIGTA502202EA 603 EUR
ORIGTA502202
Anti-MAPK7 Mouse Monoclonal Antibody [clone: OTI1A4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK7 (mitogen-activated protein kinase 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR