Anti-MTMR2 Mouse Monoclonal Antibody [clone: OTI1F10]

Proveedor: OriGene
ORIGTA502200EA 603 EUR
ORIGTA502200
Anti-MTMR2 Mouse Monoclonal Antibody [clone: OTI1F10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MTMR2 (myotubularin related protein 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR