Anti-DTNA Mouse Monoclonal Antibody [clone: OTI1B2]

Proveedor: OriGene
ORIGTA502184EA 608 EUR
ORIGTA502184
Anti-DTNA Mouse Monoclonal Antibody [clone: OTI1B2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DTNA (dystrobrevin, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR