Anti-HDAC1 Mouse Monoclonal Antibody [clone: OTI5C3]
Proveedor: OriGene
ORIGTA502142EA 603 EUR
ORIGTA502142
Anti-HDAC1 Mouse Monoclonal Antibody [clone: OTI5C3]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now